Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF2B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020365.4 | NP_065098.1 | 11, 12 | 1536 | ||
XM_017002747.1 | XP_016858236.1 | 12, 13 | 1497 | |||
GenBank mRNA | AF257077.1 | - | 11, 12 | 1399 | ||
AK022399.1 | - | 11, 12 | 1398 | |||
AK024006.1 | - | 11, 12 | 1424 | |||
AK314668.1 | - | 11, 12 | 1409 | |||
BC021017.1 | - | 11, 12 | 1387 | |||
BQ776018.1 | - | 1, 2 | 251 | |||
CR457285.1 | - | 7, 8 | 790 |