Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021090.3 | NP_066576.1 | 12, 13 | 1454 | ||
NM_153050.2 | NP_694690.1 | 12, 13 | 1454 | |||
NM_153051.2 | NP_694691.1 | 12, 13 | 1454 | |||
XM_005261803.2 | XP_005261860.1 | 13, 14 | 1572 | |||
XM_005261804.3 | XP_005261861.1 | 12, 13 | 1489 | |||
XM_005261805.2 | XP_005261862.1 | 13, 14 | 1572 | |||
XM_005261806.2 | XP_005261863.1 | 12, 13 | 1454 | |||
XM_005261808.2 | XP_005261865.1 | 13, 14 | 1572 | |||
XM_017029028.1 | XP_016884517.1 | 11, 12 | 1346 | |||
XM_017029029.1 | XP_016884518.1 | 11, 12 | 1336 | |||
XM_017029030.1 | XP_016884519.1 | 11, 12 | 1352 | |||
XM_017029031.1 | XP_016884520.1 | 11, 12 | 1340 | |||
XM_017029032.1 | XP_016884521.1 | 12, 13 | 1337 | |||
XM_017029033.1 | XP_016884522.1 | 6, 7 | 773 | |||
GenBank mRNA | AB002369.1 | - | 12, 13 | 1373 | ||
AF233436.2 | - | 9, 10 | 1135 | |||
AF233437.1 | - | 9, 10 | 1135 | |||
AF233438.1 | - | 9, 10 | 1135 | |||
AK289581.1 | - | 12, 13 | 1454 | |||
BC142713.1 | - | 12, 13 | 1410 | 40148839 | ||
BC148216.1 | - | 11, 12 | 1273 | 40082266 | ||
BC151217.1 | - | 12, 13 | 1468 | 40148098 | ||
BC152455.1 | - | 12, 13 | 1373 | 8862588 | ||
CR456525.1 | - | 10, 11 | 1150 |