Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021090.3 | NP_066576.1 | 20 | 3771 | ||
NM_153050.2 | NP_694690.1 | 20 | 3687 | |||
NM_153051.2 | NP_694691.1 | 19 | 3660 | |||
XM_005261803.2 | XP_005261860.1 | 22 | 3916 | |||
XM_005261804.3 | XP_005261861.1 | 21 | 3833 | |||
XM_005261805.2 | XP_005261862.1 | 21 | 3889 | |||
XM_005261806.2 | XP_005261863.1 | 21 | 3798 | |||
XM_005261808.2 | XP_005261865.1 | 20 | 3778 | |||
XM_017029028.1 | XP_016884517.1 | 20 | 3690 | |||
XM_017029029.1 | XP_016884518.1 | 19 | 3653 | |||
XM_017029030.1 | XP_016884519.1 | 19 | 3585 | |||
XM_017029031.1 | XP_016884520.1 | 18 | 3546 | |||
XM_017029032.1 | XP_016884521.1 | 19 | 3543 | |||
XM_017029033.1 | XP_016884522.1 | 15 | 3117 | |||
XM_017029034.1 | XP_016884523.1 | 21 | 5120 | |||
GenBank mRNA | AB002369.1 | - | 20 | 3690 | ||
AF233436.2 | - | 17 | 3368 | |||
AF233437.1 | - | 16 | 3341 | |||
AF233438.1 | - | 17 | 3452 | |||
AK289581.1 | - | 20 | 3771 | |||
BC142713.1 | - | 19 | 3616 | 40148839 | ||
BC148216.1 | - | 18 | 3479 | 40082266 | ||
BC151217.1 | - | 20 | 3785 | 40148098 | ||
BC152455.1 | - | 20 | 3690 | 8862588 | ||
CR456525.1 | - | 18 | 3467 |