Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNA1I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003406.1 | NP_001003406.1 | 27, 28 | 4683 | ||
NM_021096.3 | NP_066919.2 | 28, 29 | 4788 | |||
XM_017029035.1 | XP_016884524.1 | 18, 19 | 8058 | |||
XM_017029036.1 | XP_016884525.1 | 18, 19 | 8058 | |||
XM_017029037.1 | XP_016884526.1 | 18, 19 | 8059 | |||
GenBank mRNA | AB032946.1 | - | 9, 10 | 1192 | ||
AF129133.1 | - | 28, 29 | 4788 | |||
AF142567.1 | - | 28, 29 | 4788 | |||
AF211189.1 | - | 27, 28 | 4683 | |||
AF393329.1 | - | 27, 28 | 4683 |