Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003923.2 | NP_003914.1 | 2 | 836 | ||
GenBank mRNA | AF076292.1 | - | 2 | 836 | ||
BC065377.1 | - | 2 | 339 |