Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199279.1 | NP_001186208.1 | 4 | 1481 | ||
NM_004198.3 | NP_004189.1 | 5 | 1526 | |||
GenBank mRNA | AK298798.1 | - | 4 | 1305 | ||
AK313521.1 | - | 5 | 1372 | |||
BC014456.1 | - | 5 | 1216 | 4862198 | ||
U62435.1 | - | 5 | 1312 | |||
Y16282.1 | - | 5 | 1170 |