Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELENBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258288.1 | NP_001245217.1 | 11 | 1179 | ||
NM_001258289.1 | NP_001245218.1 | 12 | 1574 | |||
NM_003944.3 | NP_003935.2 | 12 | 1365 | |||
GenBank mRNA | AK023875.1 | - | 8 | 2456 | ||
AK296661.1 | - | 12 | 1543 | |||
AK298355.1 | - | 12 | 1230 | |||
AK303815.1 | - | 11 | 1145 | |||
AK315643.1 | - | 12 | 1335 | |||
BC009084.1 | - | 12 | 1319 | 3853674 | ||
BC032997.1 | - | 10 | 1750 | 4839079 | ||
BQ045424.1 | - | 1 | 400 | |||
CR456852.1 | - | 11 | 1272 | |||
EU794660.1 | - | 12 | 1319 |