Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BRSK2 SIRNA Assays ›
Gene Symbol:
BRSK2
Gene Name:
BR serine/threonine kinase 2
Gene Aliases:
C11orf7, PEN11B, SAD1, SADA, STK29
Chromosome Location:
Chr.11: 1389899 - 1462689 on Build GRCh38
UniGene ID:
Hs.170819
Species:
Human
siRNA ID
HSS113321
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BRSK2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256627.1
NP_001243556.1
8
1088
NM_001256629.1
NP_001243558.1
8
1088
NM_001256630.1
NP_001243559.1
8
1093
NM_001282218.1
NP_001269147.1
8
1075
NM_003957.3
NP_003948.2
8
1088
NR_046331.1
8
1088
XM_005253218.3
XP_005253275.1
8
800
XM_006718374.2
XP_006718437.1
8
800
XM_006718376.2
XP_006718439.1
8
712
XM_011520461.2
XP_011518763.1
8
712
XM_011520462.2
XP_011518764.1
8
712
XM_017018532.1
XP_016874021.1
8
712
XM_017018533.1
XP_016874022.1
8
800
XM_017018534.1
XP_016874023.1
8
712
XM_017018535.1
XP_016874024.1
8
712
XM_017018536.1
XP_016874025.1
8
800
XM_017018537.1
XP_016874026.1
8
712
XM_017018538.1
XP_016874027.1
4
289
GenBank mRNA
AF533876.1
-
8
1086
AF533877.1
-
8
1086
AF533878.1
-
8
1086
AF533879.1
-
8
1086
AF533880.1
-
8
1086
AJ006701.1
-
6
491
AK122851.1
-
8
1077
AK131534.1
-
8
1093
AK297211.1
-
4
446
AK297306.1
-
4
249
AY166857.1
-
8
1088
AY505125.1
-
8
702
DA059153.1
-
4
243
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
protein phosphorylation
exocytosis
mitotic nuclear division
axonogenesis
peptidyl-serine phosphorylation
establishment of cell polarity
neuron differentiation
actin cytoskeleton reorganization
ERAD pathway
regulation of ATPase activity
cell division
regulation of insulin secretion involved in cellular response to glucose stimulus
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
regulation of retrograde protein transport, ER to cytosol
magnesium ion binding
protein serine/threonine kinase activity
ATP binding
protein kinase binding
tau-protein kinase activity
ATPase binding
ATPase regulator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0