Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHBDL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278720.1 | NP_001265649.1 | 3 | 425 | ||
NM_001318733.1 | NP_001305662.1 | 2 | 522 | |||
XM_017023849.1 | XP_016879338.1 | 2 | 525 | |||
XM_017023850.1 | XP_016879339.1 | 2 | 525 | |||
GenBank mRNA | AJ272344.1 | - | 3 | 385 | ||
AK294136.1 | - | 3 | 369 | |||
BC120874.2 | - | 2 | 514 | 40081041 | ||
BC120875.2 | - | 2 | 515 | 40081042 | ||
BI600093.1 | - | 3 | 433 | |||
Y17108.1 | - | 2 | 522 |