Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM4SF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003963.2 | NP_003954.2 | 4 | 441 | ||
GenBank mRNA | AF027204.1 | - | 4 | 442 | ||
BC069519.1 | - | 4 | 441 | 7262226 | ||
BC093688.1 | - | 4 | 438 | 7939533 | ||
BC117163.1 | - | 4 | 438 | 40125714 | ||
CR456925.1 | - | 4 | 410 | |||
CR541798.1 | - | 4 | 410 |