Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003972.2 | NP_003963.1 | 24 | 3433 | ||
XM_011540326.1 | XP_011538628.1 | 24 | 3532 | |||
XM_011540327.2 | XP_011538629.1 | 21 | 3198 | |||
XM_011540328.2 | XP_011538630.1 | 21 | 3026 | |||
XM_011540329.2 | XP_011538631.1 | 24 | 3656 | |||
XM_017016877.1 | XP_016872366.1 | 23 | 3495 | |||
XM_017016878.1 | XP_016872367.1 | 23 | 3325 | |||
XM_017016879.1 | XP_016872368.1 | 23 | 3449 | |||
XM_017016880.1 | XP_016872369.1 | 23 | 3453 | |||
XM_017016881.1 | XP_016872370.1 | 22 | 3302 | |||
GenBank mRNA | AB208843.1 | - | 11 | 1722 | ||
AF038362.1 | - | 24 | 3452 | |||
AJ001017.2 | - | 24 | 3347 | |||
BC112201.1 | - | 24 | 3417 | 8327669 |