Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAP7 SIRNA Assays ›
Gene Symbol:
MAP7
Gene Name:
microtubule associated protein 7
Gene Aliases:
E-MAP-115, EMAP115
Chromosome Location:
Chr.6: 136342281 - 136551015 on Build GRCh38
UniGene ID:
Hs.486548
Species:
Human
siRNA ID
HSS113374
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAP7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198608.1
NP_001185537.1
9
1254
NM_001198609.1
NP_001185538.1
9
1278
NM_001198611.1
NP_001185540.1
8
1143
NM_001198614.1
NP_001185543.1
10
1257
NM_001198615.1
NP_001185544.1
9
987
NM_001198616.1
NP_001185545.1
8
1239
NM_001198617.1
NP_001185546.1
7
1068
NM_001198618.1
NP_001185547.1
9
1117
NM_001198619.1
NP_001185548.1
9
990
NM_003980.4
NP_003971.1
9
1350
XM_006715598.2
XP_006715661.1
10
1799
XM_006715599.2
XP_006715662.1
10
1568
XM_006715600.2
XP_006715663.1
10
1544
XM_006715601.3
XP_006715664.1
9
1570
XM_011536243.2
XP_011534545.1
9
2169
XM_011536244.2
XP_011534546.1
8
2078
XM_011536245.1
XP_011534547.1
8
957
XM_011536246.2
XP_011534548.1
8
2059
XM_017011471.1
XP_016866960.1
9
1664
GenBank mRNA
AJ242501.1
-
7
657
AJ242502.1
-
8
828
AK294461.1
-
8
913
AK295749.1
-
9
1006
AK296483.1
-
9
1117
AK296556.1
-
9
1116
AK311114.1
-
7
616
AK312789.1
-
9
950
AK316062.1
-
9
990
AK316529.1
-
10
1123
BC025777.1
-
9
946
5222987
X73882.1
-
9
1039
Back To Top
More Information
Panther Classification:
Molecular Function -
non-motor microtubule binding protein
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
response to osmotic stress
establishment or maintenance of cell polarity
protein localization to plasma membrane
receptor binding
structural molecule activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0