Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC13A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003984.3 | NP_003975.1 | 2, 3 | 303 | ||
NR_027384.1 | 3, 4 | 444 | ||||
XM_006722165.2 | XP_006722228.1 | 3, 4 | 352 | |||
XM_011525452.1 | XP_011523754.1 | 2, 3 | 303 | |||
GenBank mRNA | AK303909.1 | - | 3, 4 | 352 | ||
AK314684.1 | - | 2, 3 | 303 | |||
BC096276.1 | - | 3, 4 | 398 | 40006564 | ||
BC096277.1 | - | 2, 3 | 257 | 40006565 | ||
BC096278.1 | - | 2, 3 | 257 | 40006566 | ||
BC096279.1 | - | 2, 3 | 257 | 40006572 | ||
U26209.1 | - | 2, 3 | 304 |