Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297609.1 | NP_001284538.1 | 27 | 4009 | ||
NM_004672.4 | NP_004663.3 | 28 | 4033 | |||
XM_017002771.1 | XP_016858260.1 | 26 | 3414 | |||
GenBank mRNA | AB167411.1 | - | 28 | 3764 | ||
AB208805.1 | - | 27 | 3478 | |||
AF100318.1 | - | 25 | 3178 | |||
BC129950.1 | - | 20 | 2463 | |||
BC129951.1 | - | 20 | 2463 |