Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304284.1 | NP_001291213.1 | 13 | 2491 | ||
NM_004505.3 | NP_004496.2 | 5 | 1960 | |||
XM_011524050.1 | XP_011522352.1 | 9 | 2709 | |||
XM_011524051.2 | XP_011522353.1 | 11 | 1804 | |||
XM_011524052.2 | XP_011522354.1 | 10 | 1918 | |||
XM_011524053.2 | XP_011522355.1 | 11 | 1831 | |||
XM_011524054.2 | XP_011522356.1 | 11 | 1798 | |||
XM_011524055.2 | XP_011522357.1 | 9 | 1725 | |||
XM_011524056.2 | XP_011522358.1 | 6 | 1678 | |||
XM_011524057.2 | XP_011522359.1 | 10 | 1907 | |||
XM_011524058.2 | XP_011522360.1 | 10 | 1907 | |||
XM_011524059.2 | XP_011522361.1 | 10 | 1907 | |||
XM_017025290.1 | XP_016880779.1 | 8 | 2547 | |||
GenBank mRNA | AB449915.1 | - | 4 | 261 | ||
AY143550.1 | - | 5 | 1960 | |||
BX647719.1 | - | 13 | 2492 | |||
X63546.1 | - | 5 | 1957 | |||
X63547.1 | - | 5 | 1957 |