Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128610.2 | NP_001122082.1 | 15 | 2770 | ||
NM_001283049.1 | NP_001269978.1 | 12 | 2312 | |||
NM_005154.4 | NP_005145.3 | 15 | 2630 | |||
XM_006720761.3 | XP_006720824.1 | 16 | 2950 | |||
XM_006720762.3 | XP_006720825.1 | 15 | 2863 | |||
XM_011522193.2 | XP_011520495.1 | 16 | 2787 | |||
XM_017022718.1 | XP_016878207.1 | 14 | 2538 | |||
XM_017022719.1 | XP_016878208.1 | 14 | 2655 | |||
XM_017022720.1 | XP_016878209.1 | 15 | 2700 | |||
XM_017022721.1 | XP_016878210.1 | 16 | 2687 | |||
XM_017022722.1 | XP_016878211.1 | 14 | 2610 | |||
GenBank mRNA | AK292400.1 | - | 15 | 2625 | ||
AK296480.1 | - | 12 | 2302 | |||
AK300590.1 | - | 4 | 861 | |||
BC110590.2 | - | 15 | 2596 | 40027270 | ||
BX537420.1 | - | 15 | 2625 | |||
D29956.1 | - | 15 | 2744 |