Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004688.2 | NP_004679.2 | 8 | 1238 | ||
XM_005246941.2 | XP_005246998.1 | 8 | 1101 | |||
XM_017005247.1 | XP_016860736.1 | 8 | 1311 | |||
GenBank mRNA | AB451305.1 | - | 7 | 908 | ||
AK291548.1 | - | 8 | 1226 | |||
BC001268.1 | - | 8 | 958 | 3452659 | ||
BC021987.2 | - | 7 | 914 | 4096531 | ||
U32849.1 | - | 8 | 1188 |