Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004688.2 | NP_004679.2 | 3 | 487 | ||
XM_005246941.2 | XP_005246998.1 | 3 | 350 | |||
XM_017005247.1 | XP_016860736.1 | 3 | 560 | |||
GenBank mRNA | AB451305.1 | - | 2 | 157 | ||
AB451436.1 | - | 2 | 157 | |||
AK291548.1 | - | 3 | 475 | |||
AK298754.1 | - | 3 | 475 | |||
BC001268.1 | - | 3 | 207 | 3452659 | ||
BC021987.2 | - | 2 | 163 | 4096531 | ||
CB151555.1 | - | 3 | 487 | |||
EF036506.1 | - | 2 | 111 | |||
U32849.1 | - | 3 | 437 |