Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDLIM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020992.3 | NP_066272.1 | 2 | 414 | ||
GenBank mRNA | AJ310549.1 | - | 2 | 181 | ||
AK314792.1 | - | 2 | 271 | |||
BC000915.2 | - | 2 | 290 | 2985229 | ||
BC018755.1 | - | 2 | 205 | 3610938 | ||
FJ224331.1 | - | 2 | 205 | |||
U90878.1 | - | 2 | 323 |