Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNOT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271634.1 | NP_001258563.1 | 7 | 1026 | ||
NM_001271635.1 | NP_001258564.1 | 6 | 930 | |||
NM_005444.2 | NP_005435.1 | 6 | 930 | |||
NR_073390.1 | 5 | 820 | ||||
XM_011512138.2 | XP_011510440.1 | 7 | 1049 | |||
XM_017005248.1 | XP_016860737.1 | 6 | 1519 | |||
XM_017005249.1 | XP_016860738.1 | 7 | 1051 | |||
GenBank mRNA | AB500892.1 | - | 6 | 802 | ||
AK293281.1 | - | 7 | 1026 | |||
AK296037.1 | - | 2 | 83 | |||
AK316567.1 | - | 6 | 636 | |||
BC007102.1 | - | 6 | 637 | |||
BC137455.1 | - | 6 | 652 | 9021455 | ||
BC137456.1 | - | 6 | 652 | 9021456 |