Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004704.4 | NP_004695.1 | 15 | 1451 | ||
GenBank mRNA | AA465355.1 | - | 1 | 136 | ||
AJ001340.1 | - | 15 | 1411 | |||
AK313696.1 | - | 15 | 1451 | |||
BC001113.1 | - | 15 | 1434 | 3050186 | ||
BC009879.2 | - | 15 | 1416 | 3954043 | ||
BC010048.1 | - | 15 | 1434 | 3832948 | ||
BC021032.1 | - | 15 | 1434 | |||
BC023662.2 | - | 15 | 1382 | 4898690 |