Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEMF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011537317.2 | XP_011535619.1 | 3 | 497 | ||
XM_011537318.2 | XP_011535620.1 | 3 | 497 | |||
XM_017021760.1 | XP_016877249.1 | 3 | 274 | |||
GenBank mRNA | AK000913.1 | - | 3 | 228 | ||
AK293236.1 | - | 3 | 238 | |||
AK295010.1 | - | 3 | 236 | |||
AK304695.1 | - | 3 | 233 | |||
AK308537.1 | - | 3 | 233 | |||
BC006001.1 | - | 3 | 240 | |||
BC056687.1 | - | 3 | 244 | |||
BM835709.1 | - | 3 | 273 | |||
DC382921.1 | - | 3 | 522 |