Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319224.1 | NP_001306153.1 | 11 | 1870 | ||
NM_003686.4 | NP_003677.4 | 10 | 1710 | |||
NM_006027.4 | NP_006018.4 | 10 | 1710 | |||
NM_130398.3 | NP_569082.2 | 12 | 1978 | |||
XM_006711840.2 | XP_006711903.1 | 12 | 1981 | |||
XM_011544321.1 | XP_011542623.1 | 11 | 1875 | |||
XM_011544322.1 | XP_011542624.1 | 11 | 1944 | |||
XM_011544323.2 | XP_011542625.1 | 12 | 1978 | |||
XM_011544324.2 | XP_011542626.1 | 11 | 1861 | |||
XM_011544325.2 | XP_011542627.1 | 4 | 590 | |||
XM_017002793.1 | XP_016858282.1 | 10 | 1755 | |||
GenBank mRNA | AF042282.1 | - | 10 | 1614 | ||
AF060479.1 | - | 10 | 1605 | |||
AF084974.1 | - | 10 | 1605 | |||
AF091740.1 | - | 10 | 1607 | |||
AK291291.1 | - | 11 | 1858 | |||
AL080139.1 | - | 6 | 876 | |||
BC007491.2 | - | 12 | 1926 | 2967734 |