Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCSK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004716.3 | NP_004707.2 | 8 | 1659 | ||
XM_017018546.1 | XP_016874035.1 | 3 | 187 | |||
GenBank mRNA | AK097207.1 | - | 1 | 65 | ||
AK222907.1 | - | 8 | 1199 | |||
AK312429.1 | - | 8 | 1234 | |||
BC006357.2 | - | 8 | 1177 | 4125594 | ||
BC010696.1 | - | 8 | 1229 | 3868257 | ||
BT006870.1 | - | 6 | 1028 | |||
U33849.1 | - | 8 | 1179 | |||
U40623.1 | - | 6 | 1049 |