Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZW10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004724.3 | NP_004715.1 | 8 | 1113 | ||
XM_017018558.1 | XP_016874047.1 | 7 | 861 | |||
XM_017018559.1 | XP_016874048.1 | 8 | 1069 | |||
GenBank mRNA | AK303871.1 | - | 7 | 792 | ||
AK312925.1 | - | 8 | 1091 | |||
BC128264.1 | - | 8 | 1041 | 40118818 | ||
U54996.1 | - | 8 | 1039 |