Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256910.1 | NP_001243839.1 | 13 | 2212 | ||
NM_004728.3 | NP_004719.2 | 13 | 2093 | |||
XM_011540336.1 | XP_011538638.1 | 12 | 1923 | |||
XM_017016910.1 | XP_016872399.1 | 12 | 1841 | |||
GenBank mRNA | AB062394.1 | - | 5 | 572 | ||
AK025629.1 | - | 10 | 1395 | |||
AK315585.1 | - | 13 | 2042 | |||
BC008071.2 | - | 13 | 2031 | 2987980 | ||
BX648405.1 | - | 13 | 1979 | |||
CR749598.1 | - | 13 | 2213 | |||
U41387.1 | - | 13 | 2044 |