Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMYM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171162.1 | NP_001164633.1 | 19 | 3748 | ||
NM_005096.3 | NP_005087.1 | 19 | 3223 | |||
NM_201599.2 | NP_963893.1 | 19 | 3214 | |||
XM_005262309.3 | XP_005262366.1 | 19 | 3657 | |||
XM_005262310.2 | XP_005262367.1 | 19 | 3134 | |||
XM_011531062.2 | XP_011529364.1 | 19 | 3123 | |||
GenBank mRNA | AB002383.1 | - | 19 | 3122 | ||
BC069057.1 | - | 19 | 3127 | 5499547 | ||
X95808.1 | - | 20 | 3724 |