Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004760.2 | NP_004751.2 | 7 | 1196 | ||
XM_017012792.1 | XP_016868281.1 | 7 | 1008 | |||
GenBank mRNA | AB011420.1 | - | 7 | 1162 | ||
AK097170.1 | - | 2 | 1724 | |||
BC023508.2 | - | 7 | 1152 | 3638004 | ||
BC047696.1 | - | 7 | 1165 | 5761182 |
Gene Symbol: COA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005249807.4 | XP_005249864.1 | 7 | 9552 | ||
XM_006715752.2 | XP_006715815.1 | 8 | 9520 | |||
XM_017012414.1 | XP_016867903.1 | 7 | 9479 | |||
XM_017012415.1 | XP_016867904.1 | 8 | 9581 | |||
XM_017012417.1 | XP_016867906.1 | 8 | 7983 | |||
XM_017012418.1 | XP_016867907.1 | 7 | 7853 | |||
XM_017012421.1 | XP_016867910.1 | 8 | 9703 | |||
XM_017012422.1 | XP_016867911.1 | 8 | 9592 | |||
XM_017012424.1 | XP_016867913.1 | 7 | 9495 | |||
XM_017012430.1 | XP_016867919.1 | 8 | 7955 | |||
GenBank mRNA | BX537586.1 | - | 1 | 3016 |