Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYTH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004227.3 | NP_004218.1 | 8 | 725 | ||
GenBank mRNA | AJ005197.1 | - | 8 | 696 | ||
AJ223957.1 | - | 8 | 646 | |||
AK295163.1 | - | 3 | 517 | |||
AK313728.1 | - | 9 | 741 | |||
BC028717.1 | - | 8 | 761 | 4811560 |