Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYTH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292018.1 | NP_001278947.1 | 8 | 1145 | ||
NM_001292019.1 | NP_001278948.1 | 8 | 711 | |||
NM_004762.3 | NP_004753.1 | 8 | 748 | |||
NM_017456.3 | NP_059430.2 | 8 | 748 | |||
XM_011525475.2 | XP_011523777.1 | 7 | 607 | |||
XM_011525476.2 | XP_011523778.1 | 7 | 607 | |||
XM_011525477.1 | XP_011523779.1 | 6 | 526 | |||
XM_011525478.1 | XP_011523780.1 | 7 | 1067 | |||
XM_017025336.1 | XP_016880825.1 | 8 | 818 | |||
XM_017025337.1 | XP_016880826.1 | 8 | 703 | |||
XM_017025338.1 | XP_016880827.1 | 8 | 1134 | |||
GenBank mRNA | AK293894.1 | - | 8 | 1145 | ||
AK296914.1 | - | 5 | 273 | |||
AK307917.1 | - | 8 | 677 | |||
AK316075.1 | - | 8 | 704 | |||
AK316277.1 | - | 8 | 692 | |||
BC038385.1 | - | 8 | 669 | 4755203 | ||
BC050452.1 | - | 8 | 658 | 6043039 | ||
BX339155.2 | - | 8 | 728 | |||
M85169.1 | - | 8 | 698 |