Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYTH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292018.1 | NP_001278947.1 | 12 | 1428 | ||
NM_001292019.1 | NP_001278948.1 | 12 | 994 | |||
NM_004762.3 | NP_004753.1 | 12 | 1031 | |||
NM_017456.3 | NP_059430.2 | 11 | 1028 | |||
XM_011525475.2 | XP_011523777.1 | 11 | 890 | |||
XM_011525476.2 | XP_011523778.1 | 11 | 890 | |||
XM_011525477.1 | XP_011523779.1 | 10 | 809 | |||
XM_011525478.1 | XP_011523780.1 | 11 | 1350 | |||
XM_017025336.1 | XP_016880825.1 | 11 | 1098 | |||
XM_017025337.1 | XP_016880826.1 | 11 | 983 | |||
XM_017025338.1 | XP_016880827.1 | 11 | 1414 | |||
GenBank mRNA | AK293894.1 | - | 12 | 1428 | ||
AK296914.1 | - | 9 | 556 | |||
AK307917.1 | - | 12 | 960 | |||
AK316277.1 | - | 12 | 975 | |||
BC038385.1 | - | 12 | 952 | 4755203 | ||
BC050452.1 | - | 12 | 941 | 6043039 | ||
M85169.1 | - | 12 | 981 |