Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIWIL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190971.1 | NP_001177900.1 | 10 | 1342 | ||
NM_004764.4 | NP_004755.2 | 10 | 1342 | |||
XM_011539002.2 | XP_011537304.1 | 10 | 1216 | |||
XM_011539003.2 | XP_011537305.1 | 10 | 1147 | |||
XM_011539004.2 | XP_011537306.1 | 10 | 1285 | |||
XM_011539005.1 | XP_011537307.1 | 10 | 1250 | |||
XM_011539006.2 | XP_011537308.1 | 2 | 142 | |||
XM_017020229.1 | XP_016875718.1 | 10 | 1250 | |||
GenBank mRNA | AB274731.1 | - | 9 | 1071 | ||
AF104260.2 | - | 10 | 1127 | |||
AF264004.1 | - | 7 | 813 | |||
AF387507.1 | - | 10 | 1134 | |||
AK093133.1 | - | 10 | 1124 | |||
BC028581.2 | - | 10 | 1139 | 4827445 |