Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIWIL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190971.1 | NP_001177900.1 | 14 | 1899 | ||
NM_004764.4 | NP_004755.2 | 14 | 1899 | |||
XM_011539002.2 | XP_011537304.1 | 14 | 1773 | |||
XM_011539003.2 | XP_011537305.1 | 14 | 1704 | |||
XM_011539004.2 | XP_011537306.1 | 14 | 1842 | |||
XM_011539005.1 | XP_011537307.1 | 14 | 1807 | |||
XM_011539006.2 | XP_011537308.1 | 6 | 699 | |||
XM_017020229.1 | XP_016875718.1 | 14 | 1807 | |||
GenBank mRNA | AF104260.2 | - | 14 | 1684 | ||
AF264004.1 | - | 11 | 1370 | |||
AF387507.1 | - | 14 | 1691 | |||
AK093133.1 | - | 14 | 1681 | |||
BC028581.2 | - | 14 | 1696 | 4827445 |