Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004229.3 | NP_004220.2 | 12 | 1616 | ||
XM_005272701.2 | XP_005272758.1 | 12 | 1526 | |||
XM_011544000.2 | XP_011542302.1 | 13 | 1498 | |||
XM_017029962.1 | XP_016885451.1 | 13 | 1491 | |||
GenBank mRNA | AB006651.1 | - | 12 | 1595 | ||
AF104256.1 | - | 12 | 1476 | |||
AF135802.1 | - | 12 | 1549 | |||
AF304448.1 | - | 12 | 1595 | |||
AK293070.1 | - | 12 | 1794 | |||
BC098377.1 | - | 12 | 1616 | 30527948 | ||
BC132672.1 | - | 12 | 1585 | 40146694 | ||
BC132674.1 | - | 12 | 1585 | 40146696 |