Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004770.2 | NP_004761.2 | 3 | 1374 | ||
XM_017013981.1 | XP_016869470.1 | 2 | 1352 | |||
XM_017013982.1 | XP_016869471.1 | 2 | 484 | |||
GenBank mRNA | AF338730.1 | - | 3 | 1374 | ||
AF450111.1 | - | 2 | 786 | |||
U69962.1 | - | 2 | 788 |