Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004771.3 | NP_004762.2 | 4, 5 | 652 | ||
GenBank mRNA | Y12779.1 | - | 4, 5 | 652 |