Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF3C3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012086.4 | NP_036218.1 | 17, 18 | 2687 | ||
XM_005246965.4 | XP_005247022.1 | 17, 18 | 2691 | |||
GenBank mRNA | AF085850.1 | - | 2, 3 | 229 | ||
AF133123.1 | - | 17, 18 | 2623 | |||
AK025560.1 | - | 17, 18 | 2601 | |||
AK027050.1 | - | 11, 12 | 1638 | |||
AK314718.1 | - | 17, 18 | 2628 | |||
AL137453.1 | - | 14, 15 | 2029 | |||
BC015995.1 | - | 9, 10 | 1386 | |||
BC043347.1 | - | 17, 18 | 2542 | 5498609 |