Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLP2R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004246.2 | NP_004237.1 | 11 | 1706 | ||
XM_005256861.3 | XP_005256918.1 | 9 | 882 | |||
XM_011524077.2 | XP_011522379.1 | 12 | 1820 | |||
XM_017025339.1 | XP_016880828.1 | 11 | 1763 | |||
XM_017025340.1 | XP_016880829.1 | 9 | 1539 | |||
XM_017025341.1 | XP_016880830.1 | 8 | 1432 | |||
GenBank mRNA | AF105367.1 | - | 11 | 1193 | ||
AK295765.1 | - | 11 | 1085 | |||
AK314269.1 | - | 11 | 1706 | |||
BC041176.1 | - | 8 | 762 | |||
BC043390.1 | - | 9 | 901 | |||
BC096261.1 | - | 11 | 1193 | |||
BC096262.1 | - | 11 | 1193 | 40006110 | ||
BC096263.1 | - | 11 | 1189 | |||
BC096264.1 | - | 11 | 1193 |
Set Membership: |
G-protein coupled receptor |