Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFTUD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142605.1 | NP_001136077.1 | 12 | 1256 | ||
NM_001258353.1 | NP_001245282.1 | 13 | 1502 | |||
NM_001258354.1 | NP_001245283.1 | 13 | 1335 | |||
NM_004247.3 | NP_004238.3 | 13 | 1365 | |||
GenBank mRNA | AJ505017.1 | - | 9 | 739 | ||
AK126193.1 | - | 3 | 832 | |||
AK126464.1 | - | 14 | 1171 | |||
AK128148.1 | - | 13 | 1502 | |||
AK225335.1 | - | 13 | 1125 | |||
AK293108.1 | - | 13 | 1128 | |||
AK296367.1 | - | 12 | 1059 | |||
AK297392.1 | - | 13 | 1141 | |||
AK302828.1 | - | 13 | 1005 | |||
AK316098.1 | - | 12 | 1076 | |||
BC002360.2 | - | 13 | 1135 | 2960986 | ||
CR456774.1 | - | 12 | 1104 | |||
D21163.1 | - | 13 | 1164 |