Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004784.2 | NP_004775.1 | 2 | 844 | ||
XM_006714416.3 | XP_006714479.1 | 2 | 900 | |||
XM_011532415.2 | XP_011530717.1 | 2 | 812 | |||
XM_011532416.2 | XP_011530718.1 | 2 | 812 | |||
XM_017008839.1 | XP_016864328.1 | 2 | 812 | |||
XM_017008840.1 | XP_016864329.1 | 2 | 682 | |||
XM_017008842.1 | XP_016864331.1 | 1 | 200 | |||
GenBank mRNA | AF074924.1 | - | 2 | 844 | ||
AF076605.1 | - | 1 | 630 | |||
AK295439.1 | - | 2 | 695 | |||
AY358852.1 | - | 2 | 901 | |||
BC109309.1 | - | 2 | 691 | 40035856 | ||
BC109310.1 | - | 2 | 691 | 40035859 |