Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204077.1 | NP_001191006.1 | 15 | 2499 | ||
NM_004788.3 | NP_004779.2 | 15 | 2520 | |||
GenBank mRNA | AK296369.1 | - | 15 | 2446 | ||
AK302328.1 | - | 6 | 1245 | |||
AK307737.1 | - | 10 | 1696 | |||
BC111417.1 | - | 15 | 2473 | 40038776 | ||
BC112367.1 | - | 15 | 2476 | 40038767 | ||
D50916.1 | - | 15 | 2457 |