Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271754.1 | NP_001258683.1 | 5 | 863 | ||
NM_001271755.1 | NP_001258684.1 | 5 | 934 | |||
NM_001271756.1 | NP_001258685.1 | 5 | 573 | |||
NM_004791.2 | NP_004782.1 | 6 | 1081 | |||
XM_005254100.4 | XP_005254157.1 | 6 | 1039 | |||
GenBank mRNA | AB008375.1 | - | 7 | 945 | ||
AF072752.1 | - | 6 | 1059 | |||
AK095875.1 | - | 6 | 721 | |||
AK291340.1 | - | 6 | 1001 | |||
AK297744.1 | - | 5 | 789 | |||
AK298571.1 | - | 5 | 573 | |||
AK308194.1 | - | 5 | 851 | |||
BC036788.1 | - | 6 | 1002 | 5753239 |