Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004792.2 | NP_004783.2 | 13, 14 | 1378 | ||
XM_005246966.1 | XP_005247023.1 | 13, 14 | 1335 | |||
XM_005246967.1 | XP_005247024.1 | 13, 14 | 1382 | |||
GenBank mRNA | AK307552.1 | - | 10, 11 | 1160 | ||
BC073875.1 | - | 12, 13 | 1224 | |||
BC109373.1 | - | 13, 14 | 1382 | |||
BC111693.1 | - | 12, 13 | 1320 | 40034815 | ||
U40763.1 | - | 14, 15 | 1643 | |||
X99717.1 | - | 13, 14 | 1315 |