Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLAA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031689.2 | NP_001026859.1 | 12 | 2067 | ||
NM_001321546.1 | NP_001308475.1 | 11 | 2001 | |||
GenBank mRNA | AF083395.1 | - | 11 | 1471 | ||
AF145020.1 | - | 12 | 1866 | |||
AJ238243.1 | - | 11 | 1499 | |||
AK001642.1 | - | 12 | 1770 | |||
AK223544.1 | - | 11 | 1502 | |||
AL133608.1 | - | 12 | 2060 | |||
BC032551.1 | - | 12 | 1749 | 5555038 |