Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184732.1 | NP_001171661.1 | 4, 5 | 712 | ||
NM_001184733.1 | NP_001171662.1 | 4, 5 | 460 | |||
NM_001184736.1 | NP_001171665.1 | 3, 4 | 375 | |||
NM_004254.3 | NP_004245.2 | 4, 5 | 733 | |||
XM_011545364.1 | XP_011543666.1 | 3, 4 | 308 | |||
GenBank mRNA | AB042505.1 | - | 4, 5 | 715 | ||
AF097491.1 | - | 3, 4 | 588 | |||
AK298341.1 | - | 4, 5 | 430 | |||
AK298351.1 | - | 5, 6 | 685 | |||
AK307850.1 | - | 3, 4 | 340 | |||
AK313187.1 | - | 4, 5 | 706 | |||
BC022387.1 | - | 4, 5 | 727 | 4608542 | ||
BP276255.1 | - | 4, 5 | 430 |