Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRHPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012203.1 | NP_036335.1 | 9 | 1012 | ||
XM_005251631.1 | XP_005251688.1 | 6 | 705 | |||
XM_011518073.2 | XP_011516375.1 | 9 | 1386 | |||
GenBank mRNA | AF113215.1 | - | 9 | 1020 | ||
AF113251.1 | - | 9 | 1030 | |||
AF134895.1 | - | 9 | 1012 | |||
AF146018.1 | - | 9 | 1011 | |||
AK024386.1 | - | 2 | 4538 | |||
AK025978.1 | - | 9 | 1056 | |||
AK026752.1 | - | 4 | 1673 | |||
AK315690.1 | - | 9 | 1026 | |||
BC000605.2 | - | 9 | 991 | 3347232 | ||
BC003131.1 | - | 9 | 1012 | |||
BC088360.1 | - | 9 | 1010 |