Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320033.1 | NP_001306962.1 | 2 | 188 | ||
NM_004803.4 | NP_004794.2 | 1 | 233 | |||
XM_005265585.4 | XP_005265642.1 | 2 | 1010 | |||
XM_006713416.3 | XP_006713479.2 | 2 | 934 | |||
XM_006713417.3 | XP_006713480.2 | 2 | 934 | |||
XM_006713418.3 | XP_006713481.2 | 2 | 934 | |||
XM_011534245.2 | XP_011532547.1 | 2 | 1022 | |||
XM_011534247.2 | XP_011532549.1 | 2 | 934 | |||
GenBank mRNA | AB011082.1 | - | 1 | 232 | ||
AK130497.1 | - | 1 | 996 | |||
AK315317.1 | - | 2 | 186 | |||
BC075070.2 | - | 1 | 177 | 30915378 | ||
BC075071.2 | - | 1 | 177 | 30915591 | ||
BC126482.1 | - | 1 | 214 | 8992198 | ||
BC130543.1 | - | 1 | 214 | 40146574 | ||
BC144079.1 | - | 1 | 214 | |||
DB024161.1 | - | 2 | 190 |