Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004256.3 | NP_004247.2 | 2 | 471 | ||
XM_017007519.1 | XP_016863008.1 | 2 | 372 | |||
GenBank mRNA | AB010438.1 | - | 2 | 438 | ||
AK298375.1 | - | 2 | 372 | |||
AK315301.1 | - | 2 | 466 | |||
BC035973.1 | - | 2 | 466 | 4609915 | ||
DA629093.1 | - | 2 | 466 | |||
DA629311.1 | - | 2 | 471 |