Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIAO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004804.2 | NP_004795.1 | 5 | 750 | ||
GenBank mRNA | AK223257.1 | - | 5 | 671 | ||
AK299503.1 | - | 5 | 437 | |||
AK315568.1 | - | 5 | 659 | |||
BC001395.2 | - | 5 | 614 | 3050587 | ||
BC032812.1 | - | 5 | 644 | 5219740 | ||
CR456802.1 | - | 5 | 545 | |||
EF011618.1 | - | 5 | 644 | |||
U63810.1 | - | 5 | 701 |