Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGFBRAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142621.2 | NP_001136093.1 | 2 | 329 | ||
NM_004257.5 | NP_004248.2 | 2 | 332 | |||
XM_011512193.2 | XP_011510495.1 | 2 | 289 | |||
GenBank mRNA | AF022795.1 | - | 1 | 267 | ||
AK291382.1 | - | 2 | 297 | |||
BC020548.2 | - | 2 | 290 | 4420120 | ||
CT002669.1 | - | 1 | 201 | |||
DB150781.1 | - | 2 | 294 |